missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-3-(4-oxycyclohexen-1-yl)indole, 95%

Brand:  Alfa Aesar™ H63320.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15499958

  • 10855.95 SEK / 5g


Estimated Shipment date
from Supplier 23-04-2019
Add to basket

Specifications

Chemical Identifiers

MFCD28126774
4-(5-Bromo-3-indolyl)cyclohex-3-en-1-one

Specifications

5-Bromo-3-(4-oxycyclohexen-1-yl)indole
MFCD28126774
290.16
95%
5g
4-(5-Bromo-3-indolyl)cyclohex-3-en-1-one
Solid
Videos
SDS