missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-3-fluoropyridine-2-carboxylic acid, 98%

Brand:  Alfa Aesar™ H64466.MD

Additional Details : CAS Number : 669066-91-5 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15478936

  • 319.13 SEK / 250mg


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Bromo-3-fluoropyridine-2-carboxylic acid
220
11103426
669066-91-5
MFCD07437865
5-Bromo-3-fluoropicolinic acid

Specifications

5-Bromo-3-fluoropyridine-2-carboxylic acid
175° to 180°C
98%
220
11103426
669066-91-5
Solid
250mg
MFCD07437865
5-Bromo-3-fluoropicolinic acid
Videos
SDS