missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside, ≥98%, Molecular Biology Reagent, MP Biomedicals™

Catalog Number p-4774879
Click to view available options
Quantity:
10 mg
100 mg
500 mg
This item is not returnable. View return policy
7240-90-6
C14H15BrClNO6
408.629
MFCD00005666
OPIFSICVWOWJMJ-AEOCFKNESA-N
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
65181
CHEBI:75055
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
This item is not returnable. View return policy
7240-90-6
C14H15BrClNO6
408.629
MFCD00005666
OPIFSICVWOWJMJ-AEOCFKNESA-N
x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
65181
CHEBI:75055
(2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside is used as indigogenic substrate for β-galactosidase, for detection of β-galactosidase-positive clones, and the identification of lac and bacterial colonies or phage plaques

Color White
Assay Percent Range ≥98%
Quantity 10 mg
Formula Weight 408.6
Grade Molecular Biology Reagent
Physical Form Powder
Chemical Name or Material 5-Bromo-4-Chloro-3-Indoyl-β-D-Galactopyranoside, FTIR
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'