missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-4-Chloro-3-Indolyl-β-D-Glucuronide Cyclohexylammonium Salt, MP Biomedicals™
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Specifications
Melting Point | 241°C to 243°C (literature) |
Quantity | 5 mg |
Solubility Information | Soluble in DMF (2mg/mL; clear, colorless solution). |
Optical Rotation | (c=1, DMF : H2O 1:1): –90.4 db (Lit.) |
Formula Weight | 521.8g/mol |
Percent Purity | ≥95% |
Physical Form | Crystalline Powder |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'