Learn More
5-(Bromomethyl)benzo[c][1,2,5]oxadiazole, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 32863-31-3 |
| Molecular Formula | C7H5BrN2O |
| MDL Number | MFCD00174369 |
| Quantity | 1 g |
| SMILES | BrCC1=CC2=NON=C2C=C1 |
| Molecular Weight (g/mol) | 213.03 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-(Bromomethyl)benzo[c][1,2,5]oxadiazole |