Läs mer
Alfa Aesar™ 5-Bromonicotinic acid, 98%, Thermo Scientific™
Produktkod. 11440124
Change view
Kvantitet:
5 g
25 g
100 g
Förpackningsstorlek:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11440124 100 g 100g
11420124 5 g 5g
11430124 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11440124 Leverantör Alfa Aesar™ Leverantörsnummer A13589.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20826-04-4
Molekylformel C6H4BrNO2
Molekylvikt (g/mol) 202.01
MDL-nummer MFCD00009783
InChI-nyckel FQIUCPGDKPXSLL-UHFFFAOYSA-N
Synonym 5-bromonicotinic acid, 5-bromo nicotinic acid, 5-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-bromo, nicotinic acid, 5-bromo, 5-bromo-nicotinic acid, 3-bromo-5-pyridine carboxylic acid, 3-bromo pyridine-5-carboxylic acid, pubchem1211, acmc-209fev
PubChem CID 88707
IUPAC-namn 5-bromopyridine-3-carboxylic acid
LEDER OC(=O)C1=CN=CC(Br)=C1
Specifikationer
Kemiskt namn eller material 5-Bromonicotinic acid
Smältpunkt 182°C to 186°C
Kvantitet 100 g
Beilstein 115854
Formel vikt 202.01
Procent renhet 98%