missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyridine-3-carbohydrazide, 97%, Maybridge™

1471.66 SEK

Chemical Identifiers

CAS 112193-41-6
Molecular Formula C6H6BrN3O
Molecular Weight (g/mol) 216.038
MDL Number MFCD00173916
InChI Key VERXTAUONOFMLH-UHFFFAOYSA-N
Synonym 5-bromonicotinohydrazide, 3-pyridinecarboxylicacid, 5-bromo-, hydrazide, bromonicotinohydrazide, acmc-20ea1n, maybridge1_006123, 5-bromonicotinic acid hydrazide, 5-bromo-3-pyridinecarbohydrazide, 5-bromo-nicotinic acid hydrazide, 3-pyridinecarboxylicacid,5-bromo-,hydrazide, 3-pyridinecarboxylic acid, 5-bromo-, hydrazide
PubChem CID 696365
IUPAC Name 5-bromopyridine-3-carbohydrazide
SMILES C1=C(C=NC=C1Br)C(=O)NN
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10019311
SDS View Documents
Maybridge
RDP00027EA
10g Amber Glass Bottle 1471.66 SEK
10g View alternative products
Save Up to 
»
Estimated Shipment
04-11-2020
Add to basket
 
Specifications

Chemical Identifiers

112193-41-6
216.038
VERXTAUONOFMLH-UHFFFAOYSA-N
696365
C1=C(C=NC=C1Br)C(=O)NN
C6H6BrN3O
MFCD00173916
5-bromonicotinohydrazide, 3-pyridinecarboxylicacid, 5-bromo-, hydrazide, bromonicotinohydrazide, acmc-20ea1n, maybridge1_006123, 5-bromonicotinic acid hydrazide, 5-bromo-3-pyridinecarbohydrazide, 5-bromo-nicotinic acid hydrazide, 3-pyridinecarboxylicacid,5-bromo-,hydrazide, 3-pyridinecarboxylic acid, 5-bromo-, hydrazide
5-bromopyridine-3-carbohydrazide

Specifications

5-Bromopyridine-3-carbohydrazide
10g
97.0
C6H6BrN3O
5-bromonicotinohydrazide, 3-pyridinecarboxylicacid, 5-bromo-, hydrazide, bromonicotinohydrazide, acmc-20ea1n, maybridge1_006123, 5-bromonicotinic acid hydrazide, 5-bromo-3-pyridinecarbohydrazide, 5-bromo-nicotinic acid hydrazide, 3-pyridinecarboxylicacid,5-bromo-,hydrazide, 3-pyridinecarboxylic acid, 5-bromo-, hydrazide
C1=C(C=NC=C1Br)C(=O)NN
216.038
216.04
Amber Glass Bottle
112193-41-6
97%
MFCD00173916
VERXTAUONOFMLH-UHFFFAOYSA-N
5-bromopyridine-3-carbohydrazide
696365
97%