Learn More
5-Bromopyrimidin-2(1H)-one, 97%, BLD Pharm™
Product Code. 30980319
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30980319 100 g -
30973351 25 g 25g
30973444 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30980319 Supplier BLD Pharm Supplier No. BD102754100g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 38353-06-9
Molecular Formula C4H3BrN2O
MDL Number MFCD00233952
Quantity 100 g
SMILES O=C1N=CC(Br)=CN1
Molecular Weight (g/mol) 174.98
Percent Purity 97%
Chemical Name or Material 5-Bromopyrimidin-2(1H)-one