missing translation for 'onlineSavingsMsg'
Läs mer

5-Carboxyfluorescein Dipivalate N-Hydroxysuccinimide Ester, TRC

Produktkod. 30355329
Change view
Click to view available options
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30355329 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355329 Supplier Toronto Research Chemicals Supplier No. TRCC1783535MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 5-Carboxyfluorescein Dipivalate N-Hydroxysuccinimide Ester
CAS 186032-65-5
Molekylformel C35 H31 N O11
Kvantitet 5 mg
Synonym 5-Carboxyfluorescein Dipivalate NHS Ester, 5-Carboxyfluorescein Dipivalate Succinimide Ester, 3',6'-Bis(2,2-dimethyl-1-oxopropoxy)-3-oxo-spiro[isobenzofuran-1(3H),9'-[9H]xanthene]-5-carboxylic Acid 2,5-Dioxo-1-pyrrolidinyl Ester, 5-(((2,5-Dioxopyrrolidin-1-yl)oxy)carbonyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl Bis(2,2-dimethylpropanoate)
InChI formel InChI=1S/C35H31NO11/c1-33(2,3)31(41)43-19-8-11-23-25(16-19)45-26-17-20(44-32(42)34(4,5)6)9-12-24(26)35(23)22-10-7-18(15-21(22)30(40)46-35)29(39)47-36-27(37)13-14-28(36)38/h7-12,15-17H,13-14H2,1-6H3
LEDER CC(C)(C)C(=O)Oc1ccc2c(Oc3cc(OC(=O)C(C)(C)C)ccc3C24OC(=O)c5cc(ccc45)C(=O)ON6C(=O)CCC6=O)c1
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) 3',6'-bis(2,2-dimethylpropanoyloxy)-3-oxo-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate
Molekylvikt (g/mol) 641.62
Formel vikt 641.1897
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.