missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
5-Carboxyfluorescein Dipivalate N-Hydroxysuccinimide Ester, TRC
Specifications
Specifications
| Kemiskt namn eller material | 5-Carboxyfluorescein Dipivalate N-Hydroxysuccinimide Ester |
| CAS | 186032-65-5 |
| Molekylformel | C35 H31 N O11 |
| Kvantitet | 10 mg |
| Synonym | 5-Carboxyfluorescein Dipivalate NHS Ester, 5-Carboxyfluorescein Dipivalate Succinimide Ester, 3',6'-Bis(2,2-dimethyl-1-oxopropoxy)-3-oxo-spiro[isobenzofuran-1(3H),9'-[9H]xanthene]-5-carboxylic Acid 2,5-Dioxo-1-pyrrolidinyl Ester, 5-(((2,5-Dioxopyrrolidin-1-yl)oxy)carbonyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl Bis(2,2-dimethylpropanoate) |
| InChI formel | InChI=1S/C35H31NO11/c1-33(2,3)31(41)43-19-8-11-23-25(16-19)45-26-17-20(44-32(42)34(4,5)6)9-12-24(26)35(23)22-10-7-18(15-21(22)30(40)46-35)29(39)47-36-27(37)13-14-28(36)38/h7-12,15-17H,13-14H2,1-6H3 |
| LEDER | CC(C)(C)C(=O)Oc1ccc2c(Oc3cc(OC(=O)C(C)(C)C)ccc3C24OC(=O)c5cc(ccc45)C(=O)ON6C(=O)CCC6=O)c1 |
| IUPAC-namn | (2,5-dioxopyrrolidin-1-yl) 3',6'-bis(2,2-dimethylpropanoyloxy)-3-oxo-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate |
| Molekylvikt (g/mol) | 641.62 |
| Formel vikt | 641.1897 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?