Läs mer
5-Carboxyhydroxymethyluridine, TRC
Produktkod. 30359878
Change view
Kvantitet:
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30359878 10 mg -
30352639 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30359878 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17645010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Carboxyhydroxymethyluridine
CAS 934743-11-0
Molekylformel C11 H14 N2 O9
Kvantitet 10 mg
InChI formel InChI=1S/C11H14N2O9/c14-2-4-6(16)7(17)9(22-4)13-1-3(5(15)10(19)20)8(18)12-11(13)21/h1,4-7,9,14-17H,2H2,(H,19,20)(H,12,18,21)/t4-,5?,6-,7-,9-/m1/s1
LEDER OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(C(O)C(=O)O)C(=O)NC2=O
IUPAC-namn 2-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2,4-dioxopyrimidin-5-yl]-2-hydroxyacetic acid
Molekylvikt (g/mol) 318.237
Formel vikt 318.07