missing translation for 'onlineSavingsMsg'
Learn More

(5-Carboxypentyl)(triphenyl)phosphonium bromide, 97%, Thermo Scientific™

Catalog Number p-3484500
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
50889-29-7
C24H26BrO2P
457.35
MFCD00055556
JUWYRPZTZSWLCY-UHFFFAOYSA-N
5-carboxypentyl triphenylphosphonium bromide, 5-carboxypentyl triphenyl phosphonium bromide, 5-carboxypentyl triphenylphosphanium bromide, 5-carboxypentyltriphenylphosphonium bromide, phosphonium, 5-carboxypentyl triphenyl-, bromide, acmc-20apmi, pxjvnuuwrjumaq-uhfffaoysa-m, 5-carboxyamyl triphenylphosphonium bromide
2779280
5-carboxypentyl(triphenyl)phosphanium;bromide
[Br-].OC(=O)CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
50889-29-7
C24H26BrO2P
457.35
MFCD00055556
JUWYRPZTZSWLCY-UHFFFAOYSA-N
5-carboxypentyl triphenylphosphonium bromide, 5-carboxypentyl triphenyl phosphonium bromide, 5-carboxypentyl triphenylphosphanium bromide, 5-carboxypentyltriphenylphosphonium bromide, phosphonium, 5-carboxypentyl triphenyl-, bromide, acmc-20apmi, pxjvnuuwrjumaq-uhfffaoysa-m, 5-carboxyamyl triphenylphosphonium bromide
2779280
5-carboxypentyl(triphenyl)phosphanium;bromide
[Br-].OC(=O)CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 457.35
Percent Purity 97%
Chemical Name or Material (5-Carboxypentyl)(triphenyl)phosphonium bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'