missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5-Carboxypentyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used in medicine and as pharmaceutical intermediate. It is also employed as catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in medicine and as pharmaceutical intermediate. It is also employed as catalyst.
Solubility
Soluble in Chloroform and Methanol.
Notes
Hygroscopic. Store at room temperature. Incompatible with strong oxidizing agents.
It is used in medicine and as pharmaceutical intermediate. It is also employed as catalyst.
Solubility
Soluble in Chloroform and Methanol.
Notes
Hygroscopic. Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 200°C to 202°C |
Quantity | 1 g |
Solubility Information | Soluble in Chloroform and Methanol. |
Formula Weight | 457.36 |
Percent Purity | 97% |
Chemical Name or Material | (5-Carboxypentyl)triphenylphosphonium bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'