missing translation for 'onlineSavingsMsg'
Learn More

(5-Chloro-1-benzothiophen-3-yl)methylamine, 97%, Thermo Scientific™

Catalog Number p-2695155
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
71625-90-6
C9H8ClNS
197.68
MFCD01314327
VRNXLYAXYIHHHH-UHFFFAOYSA-N
5-chlorobenzo b thiophen-3-yl methanamine, 5-chloro-1-benzothiophen-3-yl methylamine, 5-chloro-1-benzothiophen-3-yl methanamine, 1-5-chloro-1-benzothiophen-3-yl methanamine, 5-chloro-benzo b thiophene-3-methanamine, 5-chlorobenzo b thiophen-3-yl methylamine, 5-chloro-1-benzothiophene-3-yl methylamine, benzo b thiophene-3-methanamine, 5-chloro, 5-chloranyl-1-benzothiophen-3-yl methanamine, c-5-chloro-benzo b thiophen-3-yl methylamine
2798782
(5-chloro-1-benzothiophen-3-yl)methanamine
C1=CC2=C(C=C1Cl)C(=CS2)CN
This item is not returnable. View return policy
71625-90-6
C9H8ClNS
197.68
MFCD01314327
VRNXLYAXYIHHHH-UHFFFAOYSA-N
5-chlorobenzo b thiophen-3-yl methanamine, 5-chloro-1-benzothiophen-3-yl methylamine, 5-chloro-1-benzothiophen-3-yl methanamine, 1-5-chloro-1-benzothiophen-3-yl methanamine, 5-chloro-benzo b thiophene-3-methanamine, 5-chlorobenzo b thiophen-3-yl methylamine, 5-chloro-1-benzothiophene-3-yl methylamine, benzo b thiophene-3-methanamine, 5-chloro, 5-chloranyl-1-benzothiophen-3-yl methanamine, c-5-chloro-benzo b thiophen-3-yl methylamine
2798782
(5-chloro-1-benzothiophen-3-yl)methanamine
C1=CC2=C(C=C1Cl)C(=CS2)CN
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 234.15
Percent Purity 97%
Chemical Name or Material (5-Chloro-1-benzothiophen-3-yl)methylamine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'