Learn More
Alfa Aesar™ 5-Chloro-2-[4-(methylthio)phenyl]benzimidazole, 95%
Brand:  Alfa Aesar™ H64540.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15498966
  • 10374.25 SEK / 5g
Estimated Shipment date
from Supplier 04-07-2018
Add to basket
Specifications
Chemical Identifiers
5-Chloro-2-[4-(methylthio)phenyl]benzimidazole
274.77
MFCD15173049
Specifications
5-Chloro-2-[4-(methylthio)phenyl]benzimidazole
Solid
274.77
5g
95%
MFCD15173049