Läs mer
5-Chloro-2-benzimidazolinone, TRC
Produktkod. 30137707
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137707 250 mg
30136119 1 g
17962505 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137707 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC203420250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2034-23-3
Molekylformel C7 H5 Cl N2 O
Molekylvikt (g/mol) 168.58
Synonym 5-Chloro-1,3-dihydro-2H-benzimidazol-2-one, 5-Chloro-1H-benzo[d]imidazol-2(3H)-one
IUPAC-namn 5-chloro-1,3-dihydrobenzimidazol-2-one
LEDER Clc1ccc2NC(=O)Nc2c1
Specifikationer
Kemiskt namn eller material 5-Chloro-1,3-dihydrobenzimidazol-2-one
Kvantitet 250 mg
InChI formel InChI=1S/C7H5ClN2O/c8-4-1-2-5-6(3-4)10-7(11)9-5/h1-3H,(H2,9,10,11)
Formel vikt 168.009