Läs mer
Alfa Aesar™ 5-Chloro-2-fluorobenzoic acid, 97%
Produktkod. 11450228
Change view
Kvantitet:
1 g
5 g
25 g
Förpackningsstorlek:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11450228 1 g 1g
11460228 5 g 5g
11384659 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11450228 Leverantör Alfa Aesar™ Leverantörsnummer B23776.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Kemiska identifierare
CAS 394-30-9
Molekylformel C7H4ClFO2
Molekylvikt (g/mol) 174.555
MDL-nummer MFCD00665762
InChI-nyckel WGAVMKXCDMQVNF-UHFFFAOYSA-N
Synonym 2-fluoro-5-chlorobenzoic acid, 5-chloro-2-fluoro-benzoic acid, 5-chloro-2-fluorobenzoicacid, benzoic acid, 5-chloro-2-fluoro, pubchem1375, acmc-1aigz, ksc223m7h, rarechem al bo 0944, 5-chloro-2-fluorobenzoic acid, benzoicacid, 5-chloro-2-fluoro
PubChem CID 2736537
IUPAC-namn 5-chloro-2-fluorobenzoic acid
LEDER C1=CC(=C(C=C1Cl)C(=O)O)F
Specifikationer
Kemiskt namn eller material 5-Chloro-2-fluorobenzoic acid
Smältpunkt 148°C to 150°C
Kvantitet 1 g
Beilstein 2614286
Formel vikt 174.56
Procent renhet 97%