Learn More
5-Chloro-2-furaldehyde, TRC
Product Code. 30360371
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30360371 50 mg
30353266 500 mg
30354329 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30360371 Supplier Toronto Research Chemicals Supplier No. TRCC38198050MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21508-19-0
Molecular Formula C5H3O2Cl
Molecular Weight (g/mol) 130.52
IUPAC Name 5-chlorofuran-2-carbaldehyde
SMILES Clc1oc(C=O)cc1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C5H3ClO2/c6-5-2-1-4(3-7)8-5/h1-3H
Formula Weight 129.9822
Chemical Name or Material 5-Chloro-2-furaldehyde