Learn More
5-Chloro-2-methoxypyridine, 97%, ACROS Organics™
Brand:  Acros Organics 453730050
Additional Details : CAS Number : 13473-01-3 Weight : 0.01000kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Quantity 5g
 View more versions of this product
Product Code. 15229866
  • 1971.51 SEK / 5g
Estimated Shipment
09-11-2020
Add to basket
Specifications
Chemical Identifiers
13473-01-3
143.57
5-chloro-2-methoxy-pyridine, pyridine, 5-chloro-2-methoxy, 2-methoxy-5-chloro pyridine, pubchem6602, 2-methoxy-5-chloropyridine, acmc-1c02r, ksc494g7j, 5-chloranyl-2-methoxy-pyridine, 5-chloro-2-methoxypyridine
5-chloro-2-methoxypyridine
C6H6ClNO
NPYYXUYLIHZYOU-UHFFFAOYSA-N
4738276
COC1=NC=C(C=C1)Cl
Specifications
5-Chloro-2-methoxypyridine
53°C
180°C to 184°C
13473-01-3
C6H6ClNO
NPYYXUYLIHZYOU-UHFFFAOYSA-N
5-chloro-2-methoxypyridine
4738276
97%
1.1930g/mL
1.193
5g
96% min. (GC)
5-chloro-2-methoxy-pyridine, pyridine, 5-chloro-2-methoxy, 2-methoxy-5-chloro pyridine, pubchem6602, 2-methoxy-5-chloropyridine, acmc-1c02r, ksc494g7j, 5-chloranyl-2-methoxy-pyridine, 5-chloro-2-methoxypyridine
COC1=NC=C(C=C1)Cl
143.57
143.57
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 5-Chloro-2-methoxypyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Documents