missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-methoxypyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Density | 1.1930g/mL |
Boiling Point | 180°C to 184°C |
Flash Point | 53°C |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Specific Gravity | 1.193 |
Formula Weight | 143.57 |
Percent Purity | 97% |
Chemical Name or Material | 5-Chloro-2-methoxypyridine |
Safety and Handling
Product Identifier
- 5-Chloro-2-methoxypyridine, 97%
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'