Läs mer
5-Chloro-2-methyl-thieno[2,3-d]-isothiazolidin-3-one-1,1-dioxide, TRC
Produktkod. 30359053
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359053 10 mg
30356055 100 mg
30360729 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30359053 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC44821010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1333316-61-2
Molekylformel C6 H4 Cl N O3 S2
Molekylvikt (g/mol) 237.684
Synonym 5-Chloro-2-methylthieno[2,3-d]isothiazol-3(2H)-one 1,1-dioxide
IUPAC-namn 5-chloro-2-methyl-1,1-dioxothieno[2,3-d][1,2]thiazol-3-one
LEDER CN1C(=O)c2sc(Cl)cc2S1(=O)=O
Specifikationer
Kemiskt namn eller material 5-Chloro-2-methyl-thieno[2,3-d]-isothiazolidin-3-one-1,1-dioxide
Kvantitet 10 mg
InChI formel InChI=1S/C6H4ClNO3S2/c1-8-6(9)5-3(13(8,10)11)2-4(7)12-5/h2H,1H3
Formel vikt 236.932