missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Chloro-2-nitro-4-(trifluoromethyl)aniline, 95%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 112°C to 114°C |
Color | Yellow |
Quantity | 250 mg |
UN Number | UN2811 |
Formula Weight | 240.57 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 5-Chloro-2-nitro-4-(trifluoromethyl)aniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'