Learn More
5-Chloro-2-nitropyridine, 97%, Thermo Scientific™
Product Code. 11421650
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421650 25 g 25g
11401650 1 g 1g
11411650 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11421650 Supplier Thermo Scientific Alfa Aesar Supplier No. H27000.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 52092-47-4
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.541
MDL Number MFCD03092916
InChI Key YUBHMOQVHOODEI-UHFFFAOYSA-N
Synonym 2-nitro-5-chloropyridine, 5-chloro-2-nitro-pyridine, pyridine, 5-chloro-2-nitro, 5-chlor-2-nitropyridin, 3-chloro-6-nitropyridine, pubchem6693, acmc-209kxq, ksc494a5l, 5-chloro-2-nitropyridine, 5-chloro-2-nitropyridine 1g
PubChem CID 259806
IUPAC Name 5-chloro-2-nitropyridine
SMILES C1=CC(=NC=C1Cl)[N+](=O)[O-]
Specifications
Melting Point 120°C to 123°C
Quantity 25 g
Formula Weight 158.54
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-nitropyridine