missing translation for 'onlineSavingsMsg'
Läs mer

5-Chloro-2-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Produktkod. 11455950
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
10 g
Förpackningsstorlek:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11455950 1 g 1g
11465950 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11455950 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H26536.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90381-07-0
Molekylformel C8H4ClF3O
Molekylvikt (g/mol) 208.564
MDL-nummer MFCD03412204
InChI-nyckel CVRLMVFONQYBOC-UHFFFAOYSA-N
Synonym 5-chloro-2-trifluoromethyl benzaldehyde, 5-chloro-2-trifluoromethyl-benzaldehyde, benzaldehyde, 5-chloro-2-trifluoromethyl, pubchem2894, ksc495m1t, 3-chloro-6-trifluoromethylbenzaldehyde
PubChem CID 17750758
IUPAC-namn 5-chloro-2-(trifluoromethyl)benzaldehyde
LEDER C1=CC(=C(C=C1Cl)C=O)C(F)(F)F

Specifications

Kemiskt namn eller material 5-Chloro-2-(trifluoromethyl)benzaldehyde
Brytningsindex 1.488
Kvantitet 1 g
Känslighet Air Sensitive
Formel vikt 208.57
Procent renhet 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.