Läs mer
5-Chloro-3-[(methylamino)sulfonyl]-2-thiophenecarboxylic Acid, TRC
Produktkod. 30364739
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364739 100 mg
30355243 25 mg
30358222 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30364739 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC703740100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70374-36-6
Molekylformel C6 H6 Cl N O4 S2
Molekylvikt (g/mol) 255.7
Synonym Lornoxicam impurity 10
IUPAC-namn 5-chloro-3-(methylsulfamoyl)thiophene-2-carboxylic acid
LEDER CNS(=O)(=O)c1cc(Cl)sc1C(=O)O
Specifikationer
Kemiskt namn eller material 5-chloro-3-(methylsulfamoyl)thiophene-2-carboxylic acid
Kvantitet 100 mg
InChI formel InChI=1S/C6H6ClNO4S2/c1-8-14(11,12)3-2-4(7)13-5(3)6(9)10/h2,8H,1H3,(H,9,10)
Formel vikt 254.9427