missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-methylbenzo[b]thiophene, 97%, ACROS Organics™

Brand:  Acros Organics 450990250

Additional Details : CAS Number : 19404-18-3 Weight : 0.02000kg


Quantity 25g

 View more versions of this product

Product Code. 15219706

  • 4595.19 SEK / 25g


Estimated Shipment
09-11-2020
Add to basket


Specifications

Chemical Identifiers

19404-18-3
182.67
5-chloro-3-methylbenzo b thiophene, 5-chloro-3-methylthianaphthene, 5-chloro-3-methyl benzo b thiophene, 5-chloro-3-methylbenzothiophene, 4-methylthio phenol, zlchem 217, pubchem7372, acmc-209exr, maybridge1_005366
5-chloro-3-methyl-1-benzothiophene
C9H7ClS
UWDQVEPXORTQFO-UHFFFAOYSA-N
317317
CC1=CSC2=C1C=C(C=C2)Cl

Specifications

5-Chloro-3-methylbenzo[b]thiophene
31.0°C to 32.0°C
19404-18-3
C9H7ClS
UWDQVEPXORTQFO-UHFFFAOYSA-N
5-chloro-3-methyl-1-benzothiophene
317317
97%
87.0°C (0.5 mmHg)
25g
96% min. (GC)
5-chloro-3-methylbenzo b thiophene, 5-chloro-3-methylthianaphthene, 5-chloro-3-methyl benzo b thiophene, 5-chloro-3-methylbenzothiophene, 4-methylthio phenol, zlchem 217, pubchem7372, acmc-209exr, maybridge1_005366
CC1=CSC2=C1C=C(C=C2)Cl
182.67
182.67
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Chloro-3-methylbenzo¢b!thiophene, 97%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents