Läs mer
5-Chloro-6-(piperidin-1-yl)pyrimidine-2,4-diamine, Mikromol™
Produktkod. 30464158
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464158 4 x 25 mg
30049906 25 mg
30052429 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30464158 Leverantör Mikromol Leverantörsnummer MM0521.0600254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Chloro-6-(piperidin-1-yl)pyrimidine-2,4-diamine, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Chloro-6-(piperidin-1-yl)pyrimidine-2,4-diamine
CAS 1784008-01-0
Analyt- eller komponentnamn 5-Chloro-6-(piperidin-1-yl)pyrimidine-2,4-diamine
Frakt skick Room Temperature
Molekylformel C9 H14 Cl N5
Certifikat och efterlevnad ISO 17025
Synonym 5-Chloro-6-(1-piperidinyl)-2,4-pyrimidinediamine
InChI formel InChI=1S/C9H14ClN5/c10-6-7(11)13-9(12)14-8(6)15-4-2-1-3-5-15/h1-5H2,(H4,11,12,13,14)
Lösningstyp Neat
LEDER Nc1nc(N)c(Cl)c(n1)N2CCCCC2
IUPAC-namn 5-chloro-6-piperidin-1-ylpyrimidine-2,4-diamine
Molekylvikt (g/mol) 227.69
Kvantitet 4 x 25 mg
Formel vikt 227.0938 g/mol