missing translation for 'onlineSavingsMsg'
Läs mer

5-Chloro-8-methylquinoline, TRC

Produktkod. 30361246
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361246 50 mg
30356257 100 mg
30361075 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361246 Supplier Toronto Research Chemicals Supplier No. TRCC38482850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78941-95-4
Molekylformel C10H8NCl
Molekylvikt (g/mol) 177.63
IUPAC-namn 5-chloro-8-methylquinoline
LEDER Cc1ccc(Cl)c2cccnc12

Specifications

Kemiskt namn eller material 5-Chloro-8-methylquinoline
Kvantitet 50 mg
InChI formel InChI=1S/C10H8ClN/c1-7-4-5-9(11)8-3-2-6-12-10(7)8/h2-6H,1H3
Formel vikt 177.0345
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.