Läs mer
5-Chloro Bifeprunox Mesylate, TRC
Produktkod. 30353327
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30353327 250 mg
1 options
Produktkod. 30353327 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC364550250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Chloro Bifeprunox Mesylate
CAS 1217042-05-1
Molekylformel C25H26ClN3O5S
Kvantitet 250 mg
Synonym 2(3H)-Benzoxazolone, 7-[4-([1,1'-biphenyl]-3-ylmethyl)-1-piperazinyl]-5-chloro-, methanesulfonate (1:1) (ACI), 5-Chloro-7-[4-[(3-phenylphenyl)methyl]piperazin-1-yl]-3H-1,3-benzoxazol-2-one methanesulfonic acid, 5-Chloro-7-[4-[(3-phenylphenyl)methyl]piperazin-1-yl]-3H-1,3-benzoxazol-2-one mesylate, 5-Chloro bifeprunox mesylate, 5-Chlorobifeprunox mesylate
InChI formel InChI=1S/C24H22ClN3O2.CH4O3S/c25-20-14-21-23(30-24(29)26-21)22(15-20)28-11-9-27(10-12-28)16-17-5-4-8-19(13-17)18-6-2-1-3-7-18;1-5(2,3)4/h1-8,13-15H,9-12,16H2,(H,26,29);1H3,(H,2,3,4)
LEDER CS(=O)(=O)O.Clc1cc2NC(=O)Oc2c(c1)N3CCN(Cc4cccc(c4)c5ccccc5)CC3
IUPAC-namn 5-chloro-7-[4-[(3-phenylphenyl)methyl]piperazin-1-yl]-3H-1,3-benzoxazol-2-one;methanesulfonic acid
Molekylvikt (g/mol) 516.01