Läs mer
5-Chloro-DL-tryptophan, TRC
Produktkod. 30366233
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366233 100 mg
30368446 50 mg
30357366 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366233 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC423220100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 154-07-4
Molekylformel C11H11ClN2O2
Molekylvikt (g/mol) 238.67
IUPAC-namn 2-amino-3-(5-chloro-1H-indol-3-yl)propanoic acid
LEDER NC(Cc1c[nH]c2ccc(Cl)cc12)C(=O)O
Specifikationer
Kemiskt namn eller material 5-Chloro-DL-tryptophan
Kvantitet 100 mg
InChI formel InChI=1S/C11H11ClN2O2/c12-7-1-2-10-8(4-7)6(5-14-10)3-9(13)11(15)16/h1-2,4-5,9,14H,3,13H2,(H,15,16)
Formel vikt 238.0509