Läs mer
5-Chloro-m-salicylic Acid, TRC
Produktkod. 30361214
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361214 25 mg
30357703 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30361214 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38005525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53984-36-4
Molekylformel C7H5ClO3
Molekylvikt (g/mol) 172.57
Synonym 3-Chloro-5-hydroxybenzoic Acid, 3-Hydroxy-5-chlorobenzoic Acid
IUPAC-namn 3-chloro-5-hydroxybenzoic acid
LEDER OC(=O)c1cc(O)cc(Cl)c1
Specifikationer
Kemiskt namn eller material 5-Chloro-m-salicylic Acid
Kvantitet 25 mg
InChI formel InChI=1S/C7H5ClO3/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3,9H,(H,10,11)
Formel vikt 171.9927