Läs mer
5-Chloro-N-methylthiophene-2-carboxamide, Mikromol™
Produktkod. 30461869
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461869 4 x 25 mg
30049771 25 mg
30051509 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461869 Leverantör Mikromol Leverantörsnummer MM3629.1100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-Chloro-N-methylthiophene-2-carboxamide, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Chloro-N-methylthiophene-2-carboxamide
CAS 97799-98-9
Analyt- eller komponentnamn 5-Chloro-N-methylthiophene-2-carboxamide
Frakt skick Room Temperature
Molekylformel C6 H6 Cl N O S
Certifikat och efterlevnad ISO 9001
Synonym 5-Chloro-N-methyl-2-thiophenecarboxamide
InChI formel InChI=1S/C6H6ClNOS/c1-8-6(9)4-2-3-5(7)10-4/h2-3H,1H3,(H,8,9)
Lösningstyp Neat
LEDER CNC(=O)c1ccc(Cl)s1
IUPAC-namn 5-chloro-N-methylthiophene-2-carboxamide
Molekylvikt (g/mol) 175.64
Kvantitet 4 x 25 mg
Formel vikt 174.9859 g/mol