Läs mer
5-Chloro-N1-phenylbenzene-1,2-diamine, TRC
Produktkod. 30353504
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353504 50 mg
30362726 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353504 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37485050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-Chloro-N1-phenylbenzene-1,2-diamine
CAS 68406-47-3
Molekylformel C12H11ClN2
Kvantitet 50 mg
Synonym 2-Amino-5-Chloro-N-phenylaniline, 2-Amino-5-chlorodiphenylamine, 4-Chloro-N'-phenylbenzene-1,2-diamine, 4-Chloro-N2-phenyl-o-phenylenediamine
InChI formel InChI=1S/C12H11ClN2/c13-9-6-7-11(14)12(8-9)15-10-4-2-1-3-5-10/h1-8,15H,14H2
LEDER NC1=CC=C(Cl)C=C1NC2=CC=CC=C2
IUPAC-namn 5-chloro-N1-phenylbenzene-1,2-diamine
Molekylvikt (g/mol) 218.68
Formel vikt 218.06