Läs mer
5-Chloroacenaphthenone, TRC
Produktkod. 30365482
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365482 500 mg
30354354 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30365482 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC367065500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 37568-51-7
Molekylformel C12H7ClO
Molekylvikt (g/mol) 202.64
Synonym 5-Chloro-1(2H)-acenaphthylenone
IUPAC-namn 5-chloro-2H-acenaphthylen-1-one
LEDER Clc1ccc2CC(=O)c3cccc1c23
Specifikationer
Kemiskt namn eller material 5-Chloroacenaphthenone
Kvantitet 500 mg
InChI formel InChI=1S/C12H7ClO/c13-10-5-4-7-6-11(14)9-3-1-2-8(10)12(7)9/h1-5H,6H2
Formel vikt 202.0185