Läs mer
5-Chloronicotinic acid, 97%
Produktkod. 11316919
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11316919 1 g 1g
11326919 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11316919 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H26804.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 22620-27-5 | C6H4ClNO2 | 157.553 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22620-27-5
Molekylformel C6H4ClNO2
Molekylvikt (g/mol) 157.553
MDL-nummer MFCD00128901
InChI-nyckel XYLPLVUYPAPCML-UHFFFAOYSA-N
Synonym 5-chloronicotinic acid, 3-pyridinecarboxylic acid, 5-chloro, 5-chloro-nicotinic acid, 5-chloro-3-pyridinecarboxylic acid, 5-chlornicotins, pubchem11089, acmc-209fxn, 3-carboxy-5-chloropyridine, ksc205g2r, 3-pyridinecarboxylicacid, 5-chloro
PubChem CID 247986
IUPAC-namn 5-chloropyridine-3-carboxylic acid
LEDER C1=C(C=NC=C1Cl)C(=O)O
Specifikationer
Kemiskt namn eller material 5-Chloronicotinic acid
Smältpunkt 169°C to 171°C
Kvantitet 1 g
Formel vikt 157.56
Procent renhet 97%