Läs mer
5-Chloropyridazin-3-amine, TRC
Produktkod. 30368722
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368722 250 mg
30356573 1 g
30365058 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30368722 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC379380250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1314978-36-3
Molekylformel C4H4ClN3
Molekylvikt (g/mol) 129.55
Synonym 3-Amino-5-chloro-pyridazine
IUPAC-namn 5-chloropyridazin-3-amine
LEDER ClC1=CN=NC(N)=C1
Specifikationer
Kemiskt namn eller material 5-Chloropyridazin-3-amine
Kvantitet 250 mg
InChI formel InChI=1S/C4H4ClN3/c5-3-1-4(6)8-7-2-3/h1-2H,(H2,6,8)
Formel vikt 129.55