Läs mer
5-(Chloropyridin-2-yl-carbamoyl)pyrazine, Mikromol™
Produktkod. 30459558
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30459558 4 x 25 mg
17943216 100 mg
17973876 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30459558 Leverantör Mikromol Leverantörsnummer MM0149.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-(Chloropyridin-2-yl-carbamoyl)pyrazine, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-(Chloropyridin-2-yl-carbamoyl)pyrazine
CAS 349125-10-6
Analyt- eller komponentnamn 5-(Chloropyridin-2-yl-carbamoyl)pyrazine
Frakt skick Room Temperature
Molekylformel C10 H7 Cl N4 O
Certifikat och efterlevnad ISO 9001
Synonym 5-(Chloropyridin-2-yl-carbamoyl)pyrazine
InChI formel InChI=1S/C10H7ClN4O/c11-7-1-2-9(14-5-7)15-10(16)8-6-12-3-4-13-8/h1-6H,(H,14,15,16)
Lösningstyp Neat
LEDER Clc1ccc(NC(=O)c2cnccn2)nc1
IUPAC-namn N-(5-chloropyridin-2-yl)pyrazine-2-carboxamide
Molekylvikt (g/mol) 234.64
Kvantitet 4 x 25 mg
Formel vikt 234.0308 g/mol