Läs mer
5-Chloroquinoline, TRC
Produktkod. 30367768
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367768 500 mg
30365466 100 mg
30352177 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30367768 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC379980500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 635-27-8
Molekylformel C9 H6 Cl N
Molekylvikt (g/mol) 163.6
IUPAC-namn 5-chloroquinoline
LEDER Clc1cccc2ncccc12
Specifikationer
Kemiskt namn eller material 5-Chloroquinoline
Kvantitet 500 mg
InChI formel InChI=1S/C9H6ClN/c10-8-4-1-5-9-7(8)3-2-6-11-9/h1-6H
Formel vikt 163.0189