Learn More
5-Chlorothiophene-2-boronic acid, 97+%, Thermo Scientific Chemicals
Catalog Number p-4844094
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
162607-18-3
C4H4BClO2S
162.39
MFCD00236030
JFCLNCVCDFUJPO-UHFFFAOYSA-N
5-chlorothiophene-2-boronic acid, 5-chloro-2-thienylboronic acid, 5-chlorothiophen-2-yl boronic acid, 5-chloro-2-thiopheneboronic acid, 5-chlorothiophen-2-boronic acid, 5-chlorothiophen-2-yl boranediol, 5-chlorothiphene 2-boronic acid, 5-chloro-2-thienyl boronic acid, boronic acid, 5-chloro-2-thienyl, 5-chlorothiophene-2-boronicacid
2734324
(5-chlorothiophen-2-yl)boronic acid
OB(O)C1=CC=C(Cl)S1
162607-18-3
C4H4BClO2S
162.39
MFCD00236030
JFCLNCVCDFUJPO-UHFFFAOYSA-N
5-chlorothiophene-2-boronic acid, 5-chloro-2-thienylboronic acid, 5-chlorothiophen-2-yl boronic acid, 5-chloro-2-thiopheneboronic acid, 5-chlorothiophen-2-boronic acid, 5-chlorothiophen-2-yl boranediol, 5-chlorothiphene 2-boronic acid, 5-chloro-2-thienyl boronic acid, boronic acid, 5-chloro-2-thienyl, 5-chlorothiophene-2-boronicacid
2734324
(5-chlorothiophen-2-yl)boronic acid
OB(O)C1=CC=C(Cl)S1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 136.0°C to 140.0°C
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 162.4
Percent Purity 97+%
Chemical Name or Material 5-Chlorothiophene-2-boronic acid
compliance-icons
Product Identifier
  • 5-Chlorothiophene-2-boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.