Läs mer
5-Chlorouridine, TRC
Produktkod. 30366394
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366394 1 g
30355516 5 g
30356706 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366394 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4241001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2880-89-9
Molekylformel C9 H11 Cl N2 O6
Molekylvikt (g/mol) 278.65
IUPAC-namn 5-chloro-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
LEDER OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(Cl)C(=O)NC2=O
Specifikationer
Kemiskt namn eller material 5-Chlorouridine
Kvantitet 1 g
InChI formel InChI=1S/C9H11ClN2O6/c10-3-1-12(9(17)11-7(3)16)8-6(15)5(14)4(2-13)18-8/h1,4-6,8,13-15H,2H2,(H,11,16,17)/t4-,5-,6-,8-/m1/s1
Formel vikt 278.0306