missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-(4-methylphenyl)-4-phenylthiazole, 97%, Thermo Scientific™

Catalog Number p-7039130
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1858255-59-0
C17H12N2S
276.357
MFCD22682956
WJJRSVUYJLIIIB-UHFFFAOYSA-N
2-4-methylphenyl-4-phenyl-1,3-thiazole-5-carbonitrile, 5-cyano-2-4-methylphenyl-4-phenylthiazole
98000061
2-(4-methylphenyl)-4-phenyl-1,3-thiazole-5-carbonitrile
CC1=CC=C(C=C1)C2=NC(=C(S2)C#N)C3=CC=CC=C3
This item is not returnable. View return policy
1858255-59-0
C17H12N2S
276.357
MFCD22682956
WJJRSVUYJLIIIB-UHFFFAOYSA-N
2-4-methylphenyl-4-phenyl-1,3-thiazole-5-carbonitrile, 5-cyano-2-4-methylphenyl-4-phenylthiazole
98000061
2-(4-methylphenyl)-4-phenyl-1,3-thiazole-5-carbonitrile
CC1=CC=C(C=C1)C2=NC(=C(S2)C#N)C3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Formula Weight 276.35
Percent Purity 97%
Chemical Name or Material 5-Cyano-2-(4-methylphenyl)-4-phenylthiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'