missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-methylpyridine, 97%, Thermo Scientific™

Catalog Number p-4813216
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3222-48-8
C7H6N2
118.14
PBLOYQAQGYUPCM-UHFFFAOYSA-N
5-cyano-2-picoline, 5-cyano-2-methylpyridine, 6-methylnicotinonitrile, 6-methyl-3-pyridinecarbonitrile, 3-cyano-6-methylpyridine, 3-pyridinecarbonitrile, 6-methyl, pubchem6695, 2-methyl-5-cyanopyridine, 6-methylnicotinonitrile #, acmc-209z1l
281995
6-methylpyridine-3-carbonitrile
CC1=NC=C(C=C1)C#N
This item is not returnable. View return policy
3222-48-8
C7H6N2
118.14
PBLOYQAQGYUPCM-UHFFFAOYSA-N
5-cyano-2-picoline, 5-cyano-2-methylpyridine, 6-methylnicotinonitrile, 6-methyl-3-pyridinecarbonitrile, 3-cyano-6-methylpyridine, 3-pyridinecarbonitrile, 6-methyl, pubchem6695, 2-methyl-5-cyanopyridine, 6-methylnicotinonitrile #, acmc-209z1l
281995
6-methylpyridine-3-carbonitrile
CC1=NC=C(C=C1)C#N
Melting Point 81°C to 86°C
Assay Percent Range 96% min. (GC)
Quantity 1 g
Formula Weight 118.14
Percent Purity 97%
Chemical Name or Material 5-Cyano-2-methylpyridine
compliance-icons
Product Identifier
  • 5-Cyano-2-methylpyridine, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'