Läs mer
5-Cyanopentanamide, TRC
Produktkod. 30368185
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368185 500 mg
30363019 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30368185 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC987620500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2304-58-7
Molekylformel C6 H10 N2 O
Molekylvikt (g/mol) 126.16
Synonym 5-Cyanovaleramide
IUPAC-namn 5-cyanopentanamide
LEDER NC(=O)CCCCC#N
Specifikationer
Kemiskt namn eller material 5-Cyanopentanamide
Kvantitet 500 mg
InChI formel InChI=1S/C6H10N2O/c7-5-3-1-2-4-6(8)9/h1-4H2,(H2,8,9)
Formel vikt 126.0793