Läs mer
5-Decyano 5-Carboxymilrinone, TRC
Produktkod. 30137529
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137529 250 mg
30012719 100 mg
30006642 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137529 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD226885250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80047-38-7
Molekylformel C12H10N2O3
Molekylvikt (g/mol) 230.22
Synonym 1,6-Dihydro-2-methyl-6-oxo-[3,4'-bipyridine]-5-carboxylic Acid, Milrinone Impurity 12
IUPAC-namn 6-methyl-2-oxo-5-pyridin-4-yl-1H-pyridine-3-carboxylic acid
LEDER CC1=C(C=C(C(=O)O)C(=O)N1)c2ccncc2
Specifikationer
Kemiskt namn eller material 5-Decyano 5-Carboxymilrinone
Kvantitet 250 mg
InChI formel InChI=1S/C12H10N2O3/c1-7-9(8-2-4-13-5-3-8)6-10(12(16)17)11(15)14-7/h2-6H,1H3,(H,14,15)(H,16,17)
Formel vikt 230.0691