Läs mer
5’-Deoxy-5-fluoro Cytidine, TRC
Produktkod. 30134172
Change view
Kvantitet:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134172 100 mg
30132677 500 mg
30004616 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134172 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD232950100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66335-38-4
Molekylformel C9 H12 F N3 O4
Molekylvikt (g/mol) 245.21
Synonym 5'-deoxy-5-fluorocytidine, 5'-DFCR, 5'-Deoxy-5-fluorocytidine, Capecitabine Imp. A (EP), Capecitabine Related Compound A (USP)
IUPAC-namn 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoropyrimidin-2-one
LEDER C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(F)C(=NC2=O)N
Specifikationer
Kemiskt namn eller material 5'-Deoxy-5-fluorocytidine
Kvantitet 100 mg
InChI formel InChI=1S/C9H12FN3O4/c1-3-5(14)6(15)8(17-3)13-2-4(10)7(11)12-9(13)16/h2-3,5-6,8,14-15H,1H3,(H2,11,12,16)/t3-,5-,6-,8-/m1/s1
Formel vikt 245.0812