Läs mer
5'-Deoxy-5-fluoro-N-[(3-methylbutoxy)carbonyl]cytidine, TRC
Produktkod. 30367010
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367010 10 mg
30367001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367010 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23535010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 162204-30-0
Molekylformel C15 H22 F N3 O6
Molekylvikt (g/mol) 359.35
Synonym Cytidine, 5'-deoxy-5-fluoro-N-[(3-methylbutoxy)carbonyl]-, Carbamic acid, [1-(5-deoxy-β-D-ribofuranosyl)-5-fluoro-1,2-dihydro-2-oxo-4-pyrimidinyl]-, 3-methylbutyl ester, 5'-Deoxy-5-fluoro-N4-(isopentyloxycarbonyl)cytidine, 3-Methylbutyl [1-(5-Deoxy-β-D-ribofuranosyl)-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl]carbamate, Capecitabine Imp. E (EP)
IUPAC-namn 3-methylbutyl N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
LEDER CC(C)CCOC(=O)NC1=NC(=O)N(C=C1F)[C@@H]2O[C@H](C)[C@@H](O)[C@H]2O
Specifikationer
Kemiskt namn eller material 5'-Deoxy-5-fluoro-N-[(3-methylbutoxy)carbonyl]cytidine
Kvantitet 10 mg
InChI formel InChI=1S/C15H22FN3O6/c1-7(2)4-5-24-15(23)18-12-9(16)6-19(14(22)17-12)13-11(21)10(20)8(3)25-13/h6-8,10-11,13,20-21H,4-5H2,1-3H3,(H,17,18,22,23)/t8-,10-,11-,13-/m1/s1
Formel vikt 359.1493