Läs mer
5’-Deoxy-5-fluorouridine, TRC
Produktkod. 30137571
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137571 50 mg
30136287 100 mg
30009712 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137571 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23572550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3094-09-5
Molekylformel C9 H11 F N2 O5
Molekylvikt (g/mol) 246.19
Synonym 5'-Deoxy-5-fluorouridine, 5-Fluoro-5'-deoxyuridine, 5-Fluorodesoxyuridine, 5'-Deoxy-5-fluorouridine, Doxifluridine, Capecitabine Imp. B (EP), Capecitabine Related Compound B (USP)
IUPAC-namn 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoropyrimidine-2,4-dione
LEDER C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(F)C(=O)NC2=O
Specifikationer
Kemiskt namn eller material 5'-Deoxy-5-fluorouridine
Kvantitet 50 mg
InChI formel InChI=1S/C9H11FN2O5/c1-3-5(13)6(14)8(17-3)12-2-4(10)7(15)11-9(12)16/h2-3,5-6,8,13-14H,1H3,(H,11,15,16)/t3-,5-,6-,8-/m1/s1
Formel vikt 246.0652