Läs mer
5-Des[2-(tert-butylamino)] Bambuterol-5-ethanol, TRC
Produktkod. 30358190
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30358190 100 mg
1 options
Produktkod. 30358190 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD288990100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5-[(1RS)-1-Hydroxyethyl]-1,3-phenylene Bis(dimethylcarbamate)
CAS 112935-93-0
Molekylformel C14 H20 N2 O5
Kvantitet 100 mg
Synonym Carbamic acid, dimethyl-, 5-(1-hydroxyethyl)-1,3-phenylene ester (9CI), 5-Des[2-(tert-butylamino)] Bambuterol-5-ethanol, 5-[(1RS)-1-Hydroxyethyl]-1,3-phenylene Bis(dimethylcarbamate)
InChI formel InChI=1S/C14H20N2O5/c1-9(17)10-6-11(20-13(18)15(2)3)8-12(7-10)21-14(19)16(4)5/h6-9,17H,1-5H3
LEDER CC(O)c1cc(OC(=O)N(C)C)cc(OC(=O)N(C)C)c1
IUPAC-namn [3-(dimethylcarbamoyloxy)-5-(1-hydroxyethyl)phenyl] N,N-dimethylcarbamate
Molekylvikt (g/mol) 296.32
Formel vikt 296.1372