Läs mer
5-Deschlorolifitegrast, TRC
Produktkod. 30179943
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179943 10 mg
30179875 5 mg
30180050 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179943 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22165010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2322380-15-2
Molekylformel C29H25ClN2O7S
Molekylvikt (g/mol) 581.04
Synonym (S)-2-(2-(Benzofuran-6-carbonyl)-5-chloro-1,2,3,4-tetrahydroisoquinoline-6-carboxamido)-3-(3-(methylsulfonyl)phenyl)propanoic Acid
IUPAC-namn (2S)-2-[[2-(1-benzofuran-6-carbonyl)-5-chloro-3,4-dihydro-1H-isoquinoline-6-carbonyl]amino]-3-(3-methylsulfonylphenyl)propanoic acid
LEDER CS(=O)(=O)c1cccc(C[C@H](NC(=O)c2ccc3CN(CCc3c2Cl)C(=O)c4ccc5ccoc5c4)C(=O)O)c1
Specifikationer
Kemiskt namn eller material 5-Deschlorolifitegrast
Kvantitet 10 mg
InChI formel InChI=1S/C29H25ClN2O7S/c1-40(37,38)21-4-2-3-17(13-21)14-24(29(35)36)31-27(33)23-8-7-20-16-32(11-9-22(20)26(23)30)28(34)19-6-5-18-10-12-39-25(18)15-19/h2-8,10,12-13,15,24H,9,11,14,16H2,1H3,(H,31,33)(H,35,36)/t24-/m0/s1
Formel vikt 580.1071