Läs mer
5-Dibenzosuberenone, TRC
Produktkod. 30365412
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365412 500 mg
30354165 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30365412 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD417335500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 5H-Dibenzo[a,d]cyclohepten-5-one (Dibenzosuberenone)
CAS 2222-33-5
Anteckningar om renhetsgrad HPLC
Molekylformel C15 H10 O
Kvantitet 500 mg
Synonym 5H-Dibenzo[a,d]cyclohepten-5-one, Dibenzosuberenone
InChI formel InChI=1S/C15H10O/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-10H
LEDER O=C1c2ccccc2C=Cc3ccccc13
IUPAC-namn dibenzo[2,1-b:1',2'-e][7]annulen-11-one
Molekylvikt (g/mol) 206.24
Formel vikt 206.0732